3-[[3,5-Bis(trifluoromethyl)phenyl]amino]-4-[[(8α,9S)-6'-methoxycinchonan-9-yl]amino]-3-cyclobutene-1,2-dione ≥97%, ee 99%

Supplier: Ambeed
Warning

A307598-100MG A307598-1G A307598-250MG
77250-906EA 57.68 USD
77250-906 77250-908 77250-910
3-[[3,5-Bis(trifluoromethyl)phenyl]amino]-4-[[(8α,9S)-6'-methoxycinchonan-9-yl]amino]-3-cyclobutene-1,2-dione ≥97%, ee 99%
3-[[3,5-Bis(trifluoromethyl)phenyl]amino]-4-[[(8α,9S)-6'-methoxycinchonan-9-yl]amino]-3-cyclobutene-1,2-dione

Precaución: for Research Use Only

Formula: C₃₂H₂₈F₆N₄O₃
MW: 630.60 g/mol
MDL Number: MFCD30802180
CAS Number: 1256245-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR