2-Sulfobenzaldehyde sodium salt 97%

Supplier: Ambeed
Warning

A307582-1000G A307582-100G A307582-25G A307582-500G
77263-150EA 53 USD
77263-150 77263-152 77263-154 77263-156
2-Sulfobenzaldehyde sodium salt 97%
2-Sulfobenzaldehyde sodium salt

Precaución: for Research Use Only

Formula: C₇H₆O₄S
MDL Number: MFCD00007478
CAS Number: 1008-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR