N-(5-Bromo-1,3,4-thiadiazol-2-yl)acetamide 97%

Supplier: Ambeed
Danger

A307343-1G A307343-250MG A307343-5G
77239-354EA 80.34 USD
77239-354 77239-356 77239-358
N-(5-Bromo-1,3,4-thiadiazol-2-yl)acetamide 97%
N-(5-Bromo-1,3,4-thiadiazol-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₄H₄BrN₃OS
MW: 222.06 g/mol
CAS Number: 55114-97-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR