3-(4-Hydroxyphenyl)hex-4-ynoic acid 97%

Supplier: Ambeed
Warning

A307048-100MG A307048-1G A307048-250MG
77250-900EA 47.38 USD
77250-900 77250-902 77250-904
3-(4-Hydroxyphenyl)hex-4-ynoic acid 97%
3-(4-Hydroxyphenyl)hex-4-ynoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₃
MW: 204.222 g/mol
MDL Number: MFCD21605732
CAS Number: 865233-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR