4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carboxylic acid)) 98%

Supplier: Ambeed
Warning

A306758-250MG A306758-25G A306758-5G
77263-116EA 33.99 USD
77263-116 77263-118 77263-120
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carboxylic acid)) 98%
4',4''',4''''',4'''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1'-biphenyl]-4-carboxylic acid))

Precaución: for Research Use Only

Formula: C₅₄H₃₆O₈
MW: 812.85 g/mol
MDL Number: MFCD30537426
CAS Number: 1610858-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR