2-Amino-5-fluoro-N-phenylbenzamide 97%

Supplier: Ambeed
Warning

A306740-1G A306740-5G
77239-340EA 27.81 USD
77239-340 77239-342
2-Amino-5-fluoro-N-phenylbenzamide 97%
2-Amino-5-fluoro-N-phenylbenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁FN₂O
MW: 230.24 g/mol
MDL Number: MFCD11101521
CAS Number: 60041-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR