Lersivirine 98%

Supplier: Ambeed
Warning

A306689-10MG A306689-1MG A306689-25MG A306689-5MG A306689-50MG
77239-332EA 78 USD
77239-332 77239-334 77239-336 77239-338 77746-589
Lersivirine 98%
Lersivirine

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₄O₂
MDL Number: MFCD16619314
CAS Number: 473921-12-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR