Eln-441958 98%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-2-(3-(9-(pyridin-4-yl)-3,9-diazaspiro[5.5]undecane-3-carbonyl)phenyl)isoindolin-1-one

A306621-100MG A306621-250MG A306621-25MG A306621-50MG
77239-324EA 523.24 USD
77239-324 77239-326 77239-328 77239-330
Eln-441958 98%
Eln-441958

Precaución: for Research Use Only

Formula: C₂₉H₂₉ClN₄O₂
MW: 501.02 g/mol
MDL Number: MFCD27665467
CAS Number: 913064-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR