3-Chloropyridine-4-carboxylic acid ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-chloroisonicotinate

A306469-1G A306469-25G A306469-5G
77277-422EA 19.57 USD
77277-422 77277-424 77277-426
3-Chloropyridine-4-carboxylic acid ethyl ester 95%
3-Chloropyridine-4-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
MDL Number: MFCD03411706
CAS Number: 211678-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR