D-(-)-Isoleucine 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R)-(-)-Isoleucine, H-D-Ile-OH

A306435-100MG A306435-1G A306435-250MG A306435-5G
77290-094EA 95 USD
77290-094 77290-096 77290-098 77290-100 77290-102
D-(-)-Isoleucine 98%
D-(-)-Isoleucine

Precaución: for Research Use Only

Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17
Boiling Pt: 235
Melting Pt: 98
Density: 1.224
MDL Number: MFCD00064221
CAS Number: 319-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR