Chloroacetamide 99%

Supplier: Ambeed
Danger

Synonyms: 2-Chloroacetamide

A306407-25G A306407-100G
77263-114EA 5.15 USD
77263-114 77748-592
Chloroacetamide 99%
Chloroacetamide

Precaución: for Research Use Only

Formula: C₂H₄ClNO
MW: 93.51 g/mol
MDL Number: MFCD00008027
CAS Number: 79-07-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR