2,5-Dichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A306362-100G A306362-1G A306362-25G
77290-088EA 262.65 USD
77290-088 77290-090 77290-092
2,5-Dichlorophenylboronic acid 98%
2,5-Dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
MDL Number: MFCD01863182
CAS Number: 135145-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR