4-Aminophenyl beta-D-Galactopyranoside 95%

Supplier: Ambeed
Danger

Synonyms: (2S,3R,4S,5R,6R)-2-(4-Aminophenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A305879-100MG A305879-1G A305879-250MG
77290-076EA 44 USD
77290-076 77290-078 77290-080
4-Aminophenyl beta-D-Galactopyranoside 95%
4-Aminophenyl beta-D-Galactopyranoside

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₆
MDL Number: MFCD00067362
CAS Number: 5094-33-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR