(±)-2-Piperazinecarboxylic acid dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Piperazine-2-carboxylic acid dihydrochloride

A305698-100G A305698-10G A305698-1G A305698-25G A305698-5G
77290-062EA 125 USD
77290-062 77290-064 77290-066 77290-068 77290-070
(±)-2-Piperazinecarboxylic acid dihydrochloride 98%
(±)-2-Piperazinecarboxylic acid dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂Cl₂N₂O₂
MW: 203.07 g/mol
MDL Number: MFCD00044803
CAS Number: 3022-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR