3,4-Dichlorophenylacetylene 97%

Supplier: Ambeed
Warning

A305667-1G A305667-250MG A305667-5G A305667-25G
77277-408EA 15 USD
77277-408 77277-410 77277-412 77748-591
3,4-Dichlorophenylacetylene 97%
3,4-Dichlorophenylacetylene

Precaución: for Research Use Only

Formula: C₈H₄Cl₂
MDL Number: MFCD07783046
CAS Number: 556112-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR