1-Methyl-3-(trifluoromethyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

A305600-1G A305600-25G A305600-5G
77239-316EA 13.39 USD
77239-316 77239-318 77239-320
1-Methyl-3-(trifluoromethyl)-1H-pyrazole 98%
1-Methyl-3-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₅F₃N₂
MW: 150.104 g/mol
MDL Number: MFCD00179867
CAS Number: 154471-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR