4-Bromo-4'-tert-butylbiphenyl 98%

Supplier: Ambeed
Warning

A305389-1G A305389-5G
77290-058EA 11 USD
77290-058 77290-060
4-Bromo-4'-tert-butylbiphenyl 98%
4-Bromo-4'-tert-butylbiphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
MDL Number: MFCD01321141
CAS Number: 162258-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR