(S)-2-(3-Methylbutanamido)pentanedioic acid 98%

Supplier: Ambeed
Warning

A305128-1G A305128-250MG
77250-868EA 117.42 USD
77250-868 77250-870
(S)-2-(3-Methylbutanamido)pentanedioic acid 98%
(S)-2-(3-Methylbutanamido)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MW: 231.24 g/mol
MDL Number: MFCD12795615
CAS Number: 80154-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR