2,2,4-Trimethyl-1,2-dihydroquinolin-6-ol 95%

Supplier: Ambeed
Warning

A305120-1G A305120-25G A305120-5G
77250-862EA 46 USD
77250-862 77250-864 77250-866
2,2,4-Trimethyl-1,2-dihydroquinolin-6-ol 95%
2,2,4-Trimethyl-1,2-dihydroquinolin-6-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO
MDL Number: MFCD00193033
CAS Number: 72107-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR