Trifluoroacetamide 97%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoroacetamide

A305077-100G A305077-10G A305077-25G A305077-500G
77250-854EA 14.42 USD
77250-854 77250-856 77250-858 77250-860
Trifluoroacetamide 97%
Trifluoroacetamide

Precaución: for Research Use Only

Formula: C₂H₂F₃NO
MW: 113.04 g/mol
MDL Number: MFCD00008008
CAS Number: 354-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR