2-Chloro-5-iodophenol 97%

Supplier: Ambeed
Warning

A304497-100G A304497-10G A304497-1G A304497-250MG A304497-25G A304497-5G
77263-088EA 674.65 USD
77263-088 77263-090 77263-092 77263-094 77263-096 77263-098
2-Chloro-5-iodophenol 97%
2-Chloro-5-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄ClIO
MW: 254.4500 g/mol
MDL Number: MFCD00672969
CAS Number: 289039-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR