BG-45 98%

Supplier: Ambeed
Warning

Synonyms: BG45, N-(2-Aminophenyl)pyrazine-2-carboxamide

A304243-10MG A304243-1MG A304243-50MG A304243-5MG
77239-296EA 19 USD
77239-296 77239-298 77239-300 77239-302
BG-45 98%
BG-45

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₄O
MDL Number: MFCD09046162
CAS Number: 926259-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR