Lanifibranor 99+%

Supplier: Ambeed
Warning

Synonyms: 4-(1-(Benzo[d]thiazol-6-ylsulphonyl)-5-chloro-1H-indol-2-yl)butanoic acid, 4-(1-(1,3-Benzothiazol-6-ylsulphonyl)-5-chloro-indol-2-yl)-butan-oic acid, IVA-337

A304202-100MG A304202-10MG A304202-1MG A304202-50MG A304202-5MG
77277-392EA 142.14 USD
77277-392 77277-394 77277-396 77277-398 77277-400
Lanifibranor 99+%
Lanifibranor

Precaución: for Research Use Only

Formula: C₁₉H₁₅ClN₂O₄S₂
MW: 434.92 g/mol
MDL Number: MFCD31619253
CAS Number: 927961-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR