2'-Hydroxyacetanilide 97%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-2-aminophenol, N-(2-Hydroxyphenyl)acetamide, 2-Acetamidophenol, Acetylcholine iodide

A304113-100G A304113-25G A304113-10G A304113-500G
77250-830EA 14 USD
77250-830 77250-832 77568-614 77746-578
2'-Hydroxyacetanilide 97%
2'-Hydroxyacetanilide

Precaución: For research use only.

Formula: C₈H₉NO₂
MDL Number: MFCD00002181
CAS Number: 614-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR