STAT5 inhibitor 98%

Supplier: Ambeed
Warning

A303941-10MG A303941-1MG A303941-25MG A303941-5MG
77277-384EA 81 USD
77277-384 77277-386 77277-388 77277-390
STAT5 inhibitor 98%
STAT5 inhibitor

Precaución: for Research Use Only

Formula: C₁₆H₁₁N₃O₃
MDL Number: MFCD00124694
CAS Number: 285986-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR