3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 97%

Supplier: Ambeed
Warning

Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate, TMB-HCl hydrate, Dihydrochloride TMB hydrate

A303812-1G A303812-250MG A303812-5G
77263-078EA 6.18 USD
77263-078 77263-080 77263-082
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate 97%
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂·2HCl·xH₂O
MW: 313.27 g/mol (anhy)
MDL Number: MFCD00150104
CAS Number: 207738-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR