2-Bromo-4,5-difluoroanisole 96%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4,5-difluoro-2-methoxybenzene

A303793-100G A303793-1G A303793-25G A303793-5G
77263-070EA 262.65 USD
77263-070 77263-072 77263-074 77263-076
2-Bromo-4,5-difluoroanisole 96%
2-Bromo-4,5-difluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
MDL Number: MFCD00070753
CAS Number: 202865-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR