4-Bromo-5-nitro-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A303660-100MG A303660-10G A303660-1G A303660-250MG A303660-5G
77290-042EA 8.24 USD
77290-042 77290-044 77290-046 77290-048 77290-050
4-Bromo-5-nitro-1H-pyrazole-3-carboxylic acid 95%
4-Bromo-5-nitro-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂BrN₃O₄
MDL Number: MFCD00464256
CAS Number: 84547-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR