N-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)-4-(4-(dimethylamino)but-2-enamido)benzamide dihydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E/Z)-THZ1 dihydrochloride
A303658-10MG
A303658-1MG
A303658-25MG
A303658-50MG
A303658-5MG
77263-060EA
105.06
USD
77263-060
77263-062
77263-064
77263-066
77263-068
N-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)-4-(4-(dimethylamino)but-2-enamido)benzamide dihydrochloride 98%
N-(3-((5-Chloro-4-(1H-indol-3-yl)pyrimidin-2-yl)amino)phenyl)-4-(4-(dimethylamino)but-2-enamido)benzamide dihydrochloride
Precaución: for Research Use Only
Formula:
C₃₁H₃₀Cl₃N₇O₂ MW: 638.97 g/mol |
CAS Number:
2095433-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...