Bromo-PEG1-acid 97%

Supplier: Ambeed
Warning

A303596-100MG A303596-250MG A303596-5G A303596-1G
77277-378EA 47 USD
77277-378 77277-380 77277-382 77593-828
Bromo-PEG1-acid 97%
Bromo-PEG1-acid

Precaución: for Research Use Only

Formula: C₅H₉BrO₃
MDL Number: MFCD22574802
CAS Number: 1393330-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR