4-[(2-furylmethyl)thio]-4-methylpentan-2-one 98%

Supplier: Ambeed
Warning

A303231-100G A303231-25G A303231-500G
77290-030EA 41 USD
77290-030 77290-032 77290-034
4-[(2-furylmethyl)thio]-4-methylpentan-2-one 98%
4-[(2-furylmethyl)thio]-4-methylpentan-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂S
MDL Number: MFCD07787593
CAS Number: 64835-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR