3-Methyl-7-propylxanthine 96%

Supplier: Ambeed
Warning

A303188-100G A303188-25G A303188-5G
77250-818EA 147 USD
77250-818 77250-820 77250-822
3-Methyl-7-propylxanthine 96%
3-Methyl-7-propylxanthine

Precaución: for Research Use Only

Formula: C9H12N4O2
MDL Number: MFCD02089337
CAS Number: 55242-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR