trans-2-Nitrocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: (E)-3-(2-Nitrophenyl)acrylic acid

A302540-100G A302540-10G A302540-25G A302540-5G
77277-356EA 166.86 USD
77277-356 77277-358 77277-360 77277-362
trans-2-Nitrocinnamic acid 98%
trans-2-Nitrocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₄
MW: 193.16 g/mol
MDL Number: MFCD00007189
CAS Number: 1013-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR