3-(((3-((Dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: BIX-02188 (Random Configuration)

A302457-10MG A302457-1MG A302457-25MG A302457-50MG A302457-5MG
77263-026EA 33.99 USD
77263-026 77263-028 77263-030 77263-032 77263-034
3-(((3-((Dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide 97%
3-(((3-((Dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxamide

Precaución: for Research Use Only

Formula: C₂₅H₂₄N₄O₂
MW: 412.493 g/mol
MDL Number: MFCD14584538
CAS Number: 1094614-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR