2,6-Dichlorocinnamic acid 99%

Supplier: Ambeed
Warning

Synonyms: 3-(2,6-Dichlorophenyl)acrylic acid, (2E)-3-(2,6-Dichlorophenyl)acrylic acid

A302453-1G A302453-25G A302453-5G
77263-020EA 6.18 USD
77263-020 77263-022 77263-024
2,6-Dichlorocinnamic acid 99%
2,6-Dichlorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆Cl₂O₂
MW: 217.05 g/mol
MDL Number: MFCD00004374
CAS Number: 5345-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR