2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-Bis(trichloromethyl)-2-(4-methoxyphenyl)-1,3,5-triazine

A302449-100G A302449-1G A302449-25G A302449-500G A302449-5G
77239-270EA 273.98 USD
77239-270 77239-272 77239-274 77239-276 77239-278
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine 98%
2-(4-Methoxyphenyl)-4,6-bis(trichloromethyl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₁₂H₇Cl₆N₃O
MW: 421.91 g/mol
MDL Number: MFCD00521486
CAS Number: 3584-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR