5-Amino-2-fluoro-4-methylbenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A302342-250MG
A302342-1G
77263-018EA
17
USD
77263-018
77616-310
5-Amino-2-fluoro-4-methylbenzonitrile 97%
5-Amino-2-fluoro-4-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₇FN₂ |
MDL Number:
MFCD28131807 CAS Number: 1426136-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...