4-Chloro-3-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-α,α,α-trifluoro-m-toluic acid

A302281-100G A302281-10G A302281-25G A302281-5G
77290-008EA 115 USD
77290-008 77290-010 77290-012 77290-014
4-Chloro-3-(trifluoromethyl)benzoic acid 97%
4-Chloro-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD01631468
CAS Number: 1737-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR