4-(Dimethylamino)but-2-enoic acid hydrochloride 98% (only trans isomers)

Supplier: Ambeed
Warning

A301766-100G A301766-10G A301766-25G A301766-5G A301766-1G
77250-796EA 589.16 USD
77250-796 77250-798 77250-800 77250-802 77746-568
4-(Dimethylamino)but-2-enoic acid hydrochloride 98% (only trans isomers)
4-(Dimethylamino)but-2-enoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.61 g/mol
MDL Number: MFCD03695466
CAS Number: 98548-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR