N-(4-(3-((5-(Dimethylamino)naphthalene)-1-sulfonamido)phenyl)thiazol-2-yl)acetamide 98%

Supplier: Ambeed
Warning

A301621-10MG A301621-1MG A301621-50MG A301621-5MG
77263-010EA 94 USD
77263-010 77263-012 77263-014 77263-016
N-(4-(3-((5-(Dimethylamino)naphthalene)-1-sulfonamido)phenyl)thiazol-2-yl)acetamide 98%
N-(4-(3-((5-(Dimethylamino)naphthalene)-1-sulfonamido)phenyl)thiazol-2-yl)acetamide

Precaución: for Research Use Only

Formula: C₂₃H₂₂N₄O₃S₂
MDL Number: MFCD30377202
CAS Number: 1609402-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR