(±)-2-Octanol 98%

Supplier: Ambeed
Warning

Synonyms: (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol

A301403-100G A301403-10G A301403-25G A301403-5G
77277-332EA 59.74 USD
77277-332 77277-334 77277-336 77277-338
(±)-2-Octanol 98%
(±)-2-Octanol

Precaución: for Research Use Only

Formula: C₈H₁₈O
MW: 130.23 g/mol
MDL Number: MFCD00004591
CAS Number: 123-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR