6-Isopropyl-7-oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3-carbonitrile hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: CPI-455 hydrochloride

A301323-10MG A301323-1MG A301323-25MG A301323-5MG
77263-002EA 42 USD
77263-002 77263-004 77263-006 77263-008
6-Isopropyl-7-oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3-carbonitrile hydrochloride 98%
6-Isopropyl-7-oxo-5-phenyl-4,7-dihydropyrazolo[1,5-a]pyrimidine-3-carbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₆H₁₅ClN₄O
CAS Number: 2095432-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR