2-Bromo-7-nitro-5H-pyrrolo[2,3-b]pyrazine 97%

Supplier: Ambeed
Warning

A301171-1G A301171-250MG
77239-242EA 261 USD
77239-242 77239-244
2-Bromo-7-nitro-5H-pyrrolo[2,3-b]pyrazine 97%
2-Bromo-7-nitro-5H-pyrrolo[2,3-b]pyrazine

Precaución: for Research Use Only

Formula: C₆H₃BrN₄O₂
MDL Number: MFCD29110946
CAS Number: 1416740-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR