2,3-Dimethoxybenzoic acid 97%

Supplier: Ambeed
Warning

A301136-1000G A301136-100G A301136-10G A301136-25G A301136-500G
77250-778EA 307.99 USD
77250-778 77250-780 77250-782 77250-784 77250-786
2,3-Dimethoxybenzoic acid 97%
2,3-Dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00002432
CAS Number: 1521-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR