4'-Fluoro-2'-methoxyacetophenone 95%

Supplier: Ambeed
Warning

A301097-10G A301097-1G A301097-25G A301097-5G
77277-308EA 42 USD
77277-308 77277-310 77277-312 77277-314
4'-Fluoro-2'-methoxyacetophenone 95%
4'-Fluoro-2'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MDL Number: MFCD00061143
CAS Number: 51788-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR