Ditetradecyl-3,3'-thiodipropionate 98%

Supplier: Ambeed
Warning

Synonyms: 3,3'-Thiodipropionic Acid Ditetradecyl Ester, 3,3'-Thiodipropionic Acid Dimyristyl Ester, Dimyristyl 3,3'-Thiodipropionate, 3,3-Thiodipropionicaciddimyristylester

A300908-100G A300908-25G A300908-500G
77277-302EA 18.54 USD
77277-302 77277-304 77277-306
Ditetradecyl-3,3'-thiodipropionate 98%
Ditetradecyl-3,3'-thiodipropionate

Precaución: for Research Use Only

Formula: C₃₄H₆₆O₄S
MW: 570.96 g/mol
MDL Number: MFCD00048462
CAS Number: 16545-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR