4-Methoxyoxindole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxyindolin-2-one, 4-Methoxy-2-indolinone

A300485-1G A300485-250MG A300485-5G
77289-968EA 224.54 USD
77289-968 77289-970 77289-972
4-Methoxyoxindole 95%
4-Methoxyoxindole

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.1733 g/mol
MDL Number: MFCD07368221
CAS Number: 7699-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR