3-Amino-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-1-Boc-pyrrolidine-3-carboxylic acid, 3-amino-1-[(tert-butoxy)carbonyl]pyrrolidine-3-carboxylic acid

A300153-100MG A300153-250MG A300153-5G
77277-290EA 16.48 USD
77277-290 77277-292 77277-294
3-Amino-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid 97%
3-Amino-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.27 g/mol
MDL Number: MFCD04115499
CAS Number: 862372-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR