3-((4,5,6,7-Tetrahydro-1H-indol-2-yl)methylene)indolin-2-one 97%

Supplier: Ambeed
Warning

A299738-100MG A299738-1G A299738-250MG
77262-978EA 12.36 USD
77262-978 77262-980 77262-982
3-((4,5,6,7-Tetrahydro-1H-indol-2-yl)methylene)indolin-2-one 97%
3-((4,5,6,7-Tetrahydro-1H-indol-2-yl)methylene)indolin-2-one

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O
MW: 264.32 g/mol
MDL Number: MFCD14586259
CAS Number: 204003-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR