(S)-4-Cyclohexyl-2-(pyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

A299458-100MG A299458-1G A299458-250MG A299458-50MG
77262-966EA 136 USD
77262-966 77262-968 77262-970 77262-972
(S)-4-Cyclohexyl-2-(pyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%
(S)-4-Cyclohexyl-2-(pyridin-2-yl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O
CAS Number: 192318-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR